2-Nitro-5-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-nitropyridine, 6-Nitro-3-picoline

A103059-10G A103059-250MG A103059-25G A103059-1G A103059-5G
103796-476EA 9 USD
103796-476 103822-178 77270-310 77270-312 77270-314 77283-032 77309-048
2-Nitro-5-picoline 98%
2-Nitro-5-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
Flash Pt: 125.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00234182
CAS Number: 1074-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR