6-Nitro-m-toluidine 97%

Supplier: Ambeed
Danger

A211123-25G A211123-10G A211123-1G A211123-5G
103818-080EA 8 USD
103818-080 103848-772 77087-176 77192-646 77303-176 77303-178
6-Nitro-m-toluidine 97%
6-Nitro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Boiling Pt: 312.4 °C (760 mm Hg)
Melting Pt: 110…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007814
CAS Number: 578-46-1
UN: 2660
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR