5-Methyl-2-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A183321-100MG A183321-250MG A183321-1G A183321-5G
103815-630EA 63 USD
103815-630 103846-192 76690-634 76690-636 77301-832 77301-834
5-Methyl-2-(trifluoromethyl)pyridine 95%
5-Methyl-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₆F₃N
Boiling Pt: 147.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07774131
CAS Number: 1620-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR