5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Cyano-5-methyl-2-(2-nitroanilino)thiophene, 5-Methyl-2-((2-nitrophenyl)amino)thiophene-3-carbonitrile
A228375-100G
A228375-10G
A228375-25G
A228375-5G
103819-394EA
6
USD
103819-394
103850-302
76722-184
76722-186
77089-090
77089-092
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile 97%
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
Precaución: for Research Use Only
Formula:
C₁₂H₉N₃O₂S MW: 259.28 g/mol Boiling Pt: 425.4 °C (760 mm Hg) Flash Pt: 211.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD06408054 CAS Number: 138564-59-7 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...