5-Methyl-1-phenylpyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-1-phenyl-1H-pyrazole-3-carboxylic acid

A232613-100MG A232613-250MG A232613-1G A232613-5G
103810-840EA 54 USD
103810-840 103850-594 76771-186 76771-188 77089-524 77089-526
5-Methyl-1-phenylpyrazole-3-carboxylic acid 97%
5-Methyl-1-phenylpyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Boiling Pt: 385.4 °C (760 mm Hg)
Melting Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00124910
CAS Number: 10199-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR