5-Methyl-1-Indanone 97%

Supplier: Ambeed
Warning

A149562-10G A149562-25G A149562-5G A149562-1G
103804-058EA 7 USD
103804-058 103842-176 76973-052 77299-732 77299-734 77317-682
5-Methyl-1-Indanone 97%
5-Methyl-1-Indanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
Boiling Pt: 262.4 °C (760 mm Hg)
Melting Pt: 69…73 °C
Flash Pt: 108.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02179276
CAS Number: 4593-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR