5-Methyl-1H-pyrrole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A180932-250MG A180932-100MG
103807-572EA 147 USD
103807-572 103845-934 76872-750 77883-373
5-Methyl-1H-pyrrole-3-carboxylic acid 95%
5-Methyl-1H-pyrrole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂
Boiling Pt: 326.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09991919
CAS Number: 100047-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR