3-Methyl-1H-pyrazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-1H-pyrazole-3-carbaldehyde

A182369-100MG A182369-10G A182369-250MG
103807-702EA 31 USD
103807-702 103846-074 77029-484 77029-486 77029-488
3-Methyl-1H-pyrazole-5-carbaldehyde 97%
3-Methyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Flash Pt: 136.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159629
CAS Number: 3273-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR