5-Methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A257510-10G A257510-250MG A257510-1G A257510-5G
103812-314EA 16 USD
103812-314 103852-180 77014-102 77014-104 77091-688 77091-690
5-Methyl-1H-indazole 97%
5-Methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Boiling Pt: 285.1 °C (760 mm Hg)
Flash Pt: 133.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01606235
CAS Number: 1776-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR