4-Methyl-1H-imidazole-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-5-imidazolecarboxaldehyde

A190774-100G A190774-10G A190774-1G A190774-5G A190774-25G
103816-330EA 9 USD
103816-330 103846-932 76769-864 76769-866 77084-758 77084-760 77302-234
4-Methyl-1H-imidazole-5-carbaldehyde 98%
4-Methyl-1H-imidazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 168…170 °C
Flash Pt: 176.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00173728
CAS Number: 68282-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR