5-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methylbenzoimidazol-2(3H)-one
A150103-250MG
A150103-1G
A150103-5G
103804-126EA
40
USD
103804-126
103842-260
76768-130
77317-766
77317-768
5-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%
5-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Precaución: for Research Use Only
Formula:
C₈H₈N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00754728 CAS Number: 5400-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...