5-Methyl-1,3,4-oxadiazol-2-amine 98%

Supplier: Ambeed
Warning

A170961-10G A170961-250MG A170961-1G A170961-25G A170961-5G
103806-568EA 14 USD
103806-568 103844-842 76769-232 76769-234 77081-964 77081-966 77081-968
5-Methyl-1,3,4-oxadiazol-2-amine 98%
5-Methyl-1,3,4-oxadiazol-2-amine

Precaución: for Research Use Only

Formula: C₃H₅N₃O
MW: 99.09 g/mol
Boiling Pt: 228.8 °C (760 mm Hg)
Melting Pt: 185…186 °C
Flash Pt: 92.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03703470
CAS Number: 52838-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR