5-Methyl-1,3,4-oxadiazol-2(3H)-one 98%

Supplier: Ambeed
Warning

A112079-10G A112079-250MG A112079-25G A112079-1G A112079-5G
103797-950EA 23 USD
103797-950 103823-784 76765-908 76765-910 76765-912 77310-592 77310-594
5-Methyl-1,3,4-oxadiazol-2(3H)-one 98%
5-Methyl-1,3,4-oxadiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C3H4N2O2
MW: 100.0761 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15143213
CAS Number: 3069-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR