5-Methoxypyridin-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-5-methoxypyridine

A136172-100MG A136172-250MG A136172-10G
103802-280EA 19 USD
103802-280 77010-154 77010-156 77683-202
5-Methoxypyridin-3-ol 97%
5-Methoxypyridin-3-ol

Precaución: For research use only.

Formula: C6H7NO2
MW: 125.1300 g/mol
Melting Pt: 147.5…148.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797672
CAS Number: 109345-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR