2-Formyl-5-methoxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde

A415348-100MG A415348-250MG A415348-1G A415348-5G
103827-292EA 29 USD
103827-292 103838-336 76773-832 76773-834 76841-914 76841-916
2-Formyl-5-methoxypyridine 97%
2-Formyl-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.1360 g/mol
Boiling Pt: 248.3 °C (760 mm Hg)
Flash Pt: 104.014 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234867
CAS Number: 22187-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR