5-Methoxyisoindoline hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A260201-1G
A260201-250MG
103812-482EA
143
USD
103812-482
103852-364
77091-942
77571-604
5-Methoxyisoindoline hydrochloride 95%
5-Methoxyisoindoline hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₂ClNO MW: 185.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09997806 CAS Number: 1159822-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...