5-Methoxyoxindole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one

A347076-1G A347076-25G A347076-5G A347076-100MG A347076-10G A347076-250MG
103825-952EA 9 USD
103825-952 103836-942 76840-010 76840-012 76840-014 76876-232 76876-234 76876-236
5-Methoxyoxindole 95%
5-Methoxyoxindole

Precaución: for Research Use Only

Formula: C9H9NO2
MW: 163.1733 g/mol
Boiling Pt: 360.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01860251
CAS Number: 7699-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR