5-Methoxyindole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A186513-100G A186513-10G A186513-250MG A186513-25G A186513-5G
103815-942EA 9 USD
103815-942 103846-524 77274-320 77274-322 77274-324 77302-034 77302-036
5-Methoxyindole-2-carboxylic acid 97%
5-Methoxyindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
Boiling Pt: 447.6 °C (760 mm Hg)
Melting Pt: 199…201 °C
Flash Pt: 224.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005614
CAS Number: 4382-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR