5-Methoxybenzofuran-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A320660-10G A320660-250MG A320660-1G A320660-5G
103825-242EA 43 USD
103825-242 103836-176 76802-898 76802-900 77306-220 77306-222
5-Methoxybenzofuran-2-carboxylic acid 98%
5-Methoxybenzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₄
MW: 192.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079771
CAS Number: 10242-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR