5-Methoxy-1,3-benzothiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-methoxy-1,3-benzothiazole

A287105-10G A287105-250MG A287105-1G
103813-734EA 25 USD
103813-734 103834-872 76772-408 76772-410 76837-414
5-Methoxy-1,3-benzothiazol-2-amine 97%
5-Methoxy-1,3-benzothiazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 347.5 °C (760 mm Hg)
Flash Pt: 163.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00464314
CAS Number: 54346-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR