5-Methoxy-6-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A100458-100MG A100458-250MG A100458-1G A100458-5G
103795-452EA 64 USD
103795-452 103821-148 77160-156 77160-158 77282-492 77282-494
5-Methoxy-6-nitro-1H-indazole 95%
5-Methoxy-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₃
MW: 193.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110272
CAS Number: 724767-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR