5-Methoxy-1-tetralone 98%
Supplier: Ambeed
|
Warning
|
A329119-10G
A329119-25G
A329119-5G
A329119-1G
103825-514EA
11
USD
103825-514
103836-468
76803-020
76839-438
76839-440
77306-390
5-Methoxy-1-tetralone 98%
5-Methoxy-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₂ Boiling Pt: 313 °C (760 mm Hg) Melting Pt: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001692 CAS Number: 33892-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...