5-Methoxy-2-pyridinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxypicolinic acid

A125587-10G A125587-250MG
103800-700EA 33 USD
103800-700 77571-528 77611-070
5-Methoxy-2-pyridinecarboxylic acid 97%
5-Methoxy-2-pyridinecarboxylic acid

Precaución: Inert atmosphere, room temperature

Formula: C7H7NO3
MW: 153.1400 g/mol
Boiling Pt: 312.8 °C (760 mm Hg)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD06800961
CAS Number: 29082-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR