6-Nitro-m-anisic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-nitrobenzoic acid

A286967-25G A286967-100G A286967-5G
103834-870EA 6 USD
103834-870 76837-412 77238-982 77305-506
6-Nitro-m-anisic acid 97%
6-Nitro-m-anisic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₅
Boiling Pt: 399.7 °C (760 mm Hg)
Melting Pt: 125…130 °C
Flash Pt: 195.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00151836
CAS Number: 1882-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR