5-Methoxy-2-methylphenol 98%

Supplier: Ambeed
Warning

A278327-100MG A278327-10G A278327-250MG A278327-1G A278327-25G A278327-5G
103813-410EA 28 USD
103813-410 103853-352 76816-328 76816-330 76816-332 77093-264 77093-266 77093-268
5-Methoxy-2-methylphenol 98%
5-Methoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.164 g/mol
Boiling Pt: 243.5±20.0 °C (760 mm Hg)
Melting Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD12405727
CAS Number: 20734-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR