5-Methoxy-2-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A203228-250MG A203228-25G
103817-450EA 36 USD
103817-450 103848-104 77152-234 77901-142
5-Methoxy-2-methylbenzaldehyde 95%
5-Methoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12547802
CAS Number: 56724-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR