5-Methoxy-o-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline

A162290-10G A162290-250MG A162290-25G A162290-5G A162290-1G
103805-600EA 9 USD
103805-600 103843-850 76973-632 76973-634 77300-582 77300-584 77319-886
5-Methoxy-o-toluidine 98%
5-Methoxy-o-toluidine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18
Boiling Pt: 253
Melting Pt: 47
Density: 1.39
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075057
CAS Number: 50868-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR