5-Methoxy-2-methyl-1H-pyrrolo[2,3-c]pyridine 95%
Supplier: Ambeed
|
Warning
|
A266255-100MG
A266255-250MG
A266255-1G
103812-792EA
202
USD
103812-792
77113-816
77113-818
77304-952
5-Methoxy-2-methyl-1H-pyrrolo[2,3-c]pyridine 95%
5-Methoxy-2-methyl-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂O MW: 162.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12400733 CAS Number: 1097839-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...