5-Methoxy-1H-indazole 95%

Supplier: Ambeed
Warning

A324885-1G A324885-25G A324885-5G A324885-250MG
103825-382EA 16 USD
103825-382 103836-324 76839-244 76839-246 76839-248 77154-590
5-Methoxy-1H-indazole 95%
5-Methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 312.5 °C (760 mm Hg)
Flash Pt: 114.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07781657
CAS Number: 94444-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR