5-Methoxy-2-benzimidazolinone 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-1H-benzo[d]imidazol-2(3H)-one, 2-Hydroxy-5-methoxybenzimidazole

A120315-10G A120315-250MG A120315-1G A120315-5G A120315-100MG
103799-916EA 26 USD
103799-916 76766-416 76766-418 77297-752 77297-754 77623-996
5-Methoxy-2-benzimidazolinone 95%
5-Methoxy-2-benzimidazolinone

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
Melting Pt: 255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051946
CAS Number: 2080-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR