5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A129167-100MG
A129167-250MG
A129167-1G
103801-238EA
92
USD
103801-238
77094-906
77094-908
77313-602
5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride 95%
5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO·ClH MW: 199.677 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09026772 CAS Number: 1073968-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...