5-Isopropyl-1H-indole-2,3-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5-Isopropylindoline-2,3-dione

A499656-100MG A499656-250MG A499656-5G
103828-280EA 75 USD
103828-280 103839-368 76774-482 76774-484 77307-610
5-Isopropyl-1H-indole-2,3-dione 95%
5-Isopropyl-1H-indole-2,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02947184
CAS Number: 150560-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR