5-Isopropyl-1H-pyrazole-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A108964-250MG
A108964-1G
A108964-5G
103797-416EA
12
USD
103797-416
103823-204
77148-614
77310-034
77310-036
5-Isopropyl-1H-pyrazole-3-carboxylic acid 97%
5-Isopropyl-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₂ MW: 154.167 g/mol Melting Pt: 153…157 °C Flash Pt: 170.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD05170240 CAS Number: 92933-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...