5-Iodouridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione

A115349-100G A115349-10G A115349-250MG A115349-1G A115349-5G A115349-25G
103798-552EA 15 USD
103798-552 103824-406 77245-872 77245-874 77245-876 77311-222 77311-224 77745-852
5-Iodouridine 98%
5-Iodouridine

Precaución: for Research Use Only

Formula: C₉H₁₁IN₂O₆
MW: 370.10 g/mol
Melting Pt: 205…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006532
CAS Number: 1024-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR