5-Iodoquinolin-8-amine 95%
Supplier: Ambeed
|
Warning
|
A208887-100MG
A208887-250MG
A208887-1G
103817-894EA
125
USD
103817-894
103848-582
77012-890
77012-892
77086-914
5-Iodoquinolin-8-amine 95%
5-Iodoquinolin-8-amine
Precaución: for Research Use Only
Formula:
C₉H₇IN₂ MW: 270.07 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24160460 CAS Number: 142340-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...