5-Iodopyrimidine 98%

Supplier: Ambeed
Warning

A308425-100MG A308425-10G A308425-250MG A308425-5G
103814-560EA 28 USD
103814-560 103835-736 76787-884 76787-886 76787-888 76838-496
5-Iodopyrimidine 98%
5-Iodopyrimidine

Precaución: for Research Use Only

Formula: C4H3IN2
MW: 205.9845 g/mol
Boiling Pt: 231.6 °C (760 mm Hg)
Flash Pt: 93.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233961
CAS Number: 31462-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR