2-Amino-5-iodopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

A107127-100G A107127-10G A107127-1G A107127-25G A107127-500G A107127-5G
103822-888EA 6 USD
103822-888 77008-674 77008-676 77008-678 77008-680 77008-682 77309-756
2-Amino-5-iodopyridine 97%
2-Amino-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
Boiling Pt: 224.9 °C (760 mm Hg)
Melting Pt: 59…61 °C
Flash Pt: 131.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR