5-Iodo-N,N-dimethylpyridin-2-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Dimethylamino-5-iodopyridine
A229417-250MG
A229417-1G
103819-478EA
176
USD
103819-478
103850-378
76987-284
77303-850
5-Iodo-N,N-dimethylpyridin-2-amine 97%
5-Iodo-N,N-dimethylpyridin-2-amine
Precaución: for Research Use Only
Formula:
C₇H₉IN₂ MW: 248.06 g/mol Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD18207670 CAS Number: 380381-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...