5-Iodo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A319647-100MG A319647-250MG A319647-1G
103825-212EA 89 USD
103825-212 103836-144 76817-028 76817-030 76838-994
5-Iodo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 95%
5-Iodo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₇H₇IN₄
MW: 274.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21603870
CAS Number: 862729-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR