5-Iodo-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A105047-250MG A105047-1G A105047-5G
103796-824EA 69 USD
103796-824 103822-552 76869-380 77309-418 77309-420
5-Iodo-7H-pyrrolo[2,3-d]pyrimidine 98%
5-Iodo-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
Storage Temperature: Refrigerator
MDL Number: MFCD13189362
CAS Number: 175791-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR