4-Amino-5-iodopyrrolo[2,3-d]pyrimidine 95%

Supplier: Ambeed
Warning

A131803-250MG A131803-1G A131803-5G
103801-634EA 34 USD
103801-634 77149-642 77298-508 77314-160
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine 95%
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₅IN₄
Boiling Pt: 254.0±50.0 °C (760 mm Hg)
Flash Pt: 107.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13619882
CAS Number: 163622-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR