2-Methyl-5-iodopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-2-methylpyridine

A322953-10G A322953-1G A322953-5G
103825-316EA 44 USD
103825-316 103836-254 77277-732 77306-280 77306-282
2-Methyl-5-iodopyridine 97%
2-Methyl-5-iodopyridine

Precaución: for Research Use Only

Formula: C₆H₆IN
MW: 219.023 g/mol
Boiling Pt: 218.6 °C (760 mm Hg)
Melting Pt: 48…49 °C
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07437990
CAS Number: 695-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR