5-Iodo-2,4-dimethoxypyrimidine 95%

Supplier: Ambeed
Warning

A119284-250MG A119284-1G A119284-25G
103799-744EA 14 USD
103799-744 77108-462 77311-864 77614-930
5-Iodo-2,4-dimethoxypyrimidine 95%
5-Iodo-2,4-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇IN₂O₂
Boiling Pt: 328.7 °C (760 mm Hg)
Melting Pt: 71…75 °C
Storage Temperature: Freezer
MDL Number: MFCD00090865
CAS Number: 52522-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR