5-Iodo-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A227888-100MG A227888-250MG A227888-1G
103819-346EA 92 USD
103819-346 103850-254 76786-168 76786-170 77303-806
5-Iodo-2-(trifluoromethyl)pyridine 98%
5-Iodo-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃F₃IN
Melting Pt: 74…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11226772
CAS Number: 873107-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR