5-Iodo-1-methylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A417117-1G A417117-5G
103827-330EA 76 USD
103827-330 103838-376 77307-184 77307-186
5-Iodo-1-methylpyridin-2(1H)-one 95%
5-Iodo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23768670
CAS Number: 60154-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR