5-Iodo-1-methyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A524047-100G A524047-250MG A524047-1G A524047-5G A524047-25G A524047-10G
103828-524EA 10 USD
103828-524 103839-640 76790-106 76790-108 76843-650 76843-652 77307-720 77594-628
5-Iodo-1-methyl-1H-pyrazole 97%
5-Iodo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
Melting Pt: 76…81 °C
Flash Pt: 91.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109327
CAS Number: 34091-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR