5-Iodoindole 98%

Supplier: Ambeed
Warning

A128962-100G A128962-10G A128962-250MG A128962-1G A128962-25G A128962-5G
103801-208EA 6 USD
103801-208 77234-394 77234-396 77234-398 77313-564 77313-566 77313-568
5-Iodoindole 98%
5-Iodoindole

Precaución: for Research Use Only

Formula: C₈H₆IN
Boiling Pt: 341.7 °C (760 mm Hg)
Melting Pt: 101…104 °C
Flash Pt: 160.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00220065
CAS Number: 16066-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR