5-Iodo-1,2,3-trimethoxybenzene 95%

Supplier: Ambeed
Warning

A155143-100MG A155143-250MG A155143-25G A155143-1G
103804-720EA 19 USD
103804-720 103842-916 76768-314 76768-316 76768-318 77318-644
5-Iodo-1,2,3-trimethoxybenzene 95%
5-Iodo-1,2,3-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₃
Boiling Pt: 312.6 °C (760 mm Hg)
Flash Pt: 142.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01318153
CAS Number: 25245-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR