5-Hydroxypicolinic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-hydroxypyridine-2-carboxylic acid
A128236-100G
A128236-10G
A128236-250MG
A128236-500G
A128236-1G
A128236-25G
A128236-5G
103801-094EA
9
USD
103801-094
77285-190
77285-192
77285-194
77285-196
77313-386
77313-388
77313-390
5-Hydroxypicolinic acid 97%
5-Hydroxypicolinic acid
Precaución: for Research Use Only
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 616.2 °C (760 mm Hg) Melting Pt: 260…264 °C Flash Pt: 326.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00661309 CAS Number: 15069-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...