5-Hydroxypicolinaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxypyridine-2-carboxaldehyde

A423582-100MG A423582-10G A423582-250MG A423582-1G A423582-25G A423582-5G
103827-438EA 15 USD
103827-438 103838-488 76789-148 76789-150 76789-152 76842-122 76842-124 76842-126
5-Hydroxypicolinaldehyde 98%
5-Hydroxypicolinaldehyde

Precaución: for Research Use Only

Formula: C6H5NO2
MW: 123.1094 g/mol
Boiling Pt: 363.5 °C (760 mm Hg)
Flash Pt: 173.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697538
CAS Number: 31191-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR